Assay Details
Target Gene Details
Entrez Gene ID: | 77605 |
Gene Name: | H2A histone family, member V |
Gene Aliases: |
C530002L11Rik, H2a.z-2, H2av |
Location: |
Chr.11:6427226-6444443 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H2afv | NM_029938.1 | NP_084214.1 | ||
AK021267.1 | BAB32354.1 | |||
AK155105.1 | BAE33050.1 | |||
AK168110.1 | BAE40081.1 | |||
BC028539.1 | AAH28539.1 | |||
BC147293.1 | ||||
BC147294.1 | ||||
BC147741.1 | ||||
BC147751.1 | ||||
CJ069840.1 |
More Information
Set Membership: |
Intragenic Non-exonic |