Assay Details
Target Gene Details
Entrez Gene ID: | 399609 |
Gene Name: | RIKEN cDNA C130046K22 gene |
Gene Aliases: |
- |
Location: |
Chr.11:103697724-103725573 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C130046K22Rik | NR_102388.1 | |||
NR_102389.1 | ||||
NR_102396.1 | ||||
AK165205.1 | ||||
BY732345.1 |
Target Gene Details
Entrez Gene ID: | 56494 |
Gene Name: | golgi SNAP receptor complex member 2 |
Gene Aliases: |
2310032N09Rik, C76855, Gs27, SNARE, membrin |
Location: |
Chr.11:103676849-103697738 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gosr2 | NM_019650.3 | NP_062624.2 | ||
AF007550.1 | AAB82653.1 | |||
AK002556.1 | ||||
AK005697.1 | BAB24194.1 | |||
AK013052.1 | BAB28622.1 | |||
AK150004.1 | BAE29231.1 | |||
BC008525.1 | AAH08525.1 | |||
BC051253.1 | AAH51253.1 |
More Information
Set Membership: |
Intragenic Non-exonic |