Assay Details
Target Gene Details
Entrez Gene ID: | 103534 |
Gene Name: | mannoside acetylglucosaminyltransferase 4, isoenzyme B |
Gene Aliases: |
AA407995, GnTIVb |
Location: |
Chr.11:50225335-50235103 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mgat4b | NM_145926.3 | 3 | 656 | NP_666038.3 |
AB053218.1 | 3 | 358 | BAC55019.1 | |
AK155336.1 | 3 | 657 | BAE33202.1 | |
AK202433.1 | ||||
AK209484.1 | ||||
BY729920.1 |
More Information
Set Membership: |
Intragenic Exonic |