Assay Details
Target Gene Details
Entrez Gene ID: | 103534 |
Gene Name: | mannoside acetylglucosaminyltransferase 4, isoenzyme B |
Gene Aliases: |
AA407995, GnTIVb |
Location: |
Chr.11:50225335-50235103 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mgat4b | NM_145926.3 | NP_666038.3 | ||
AB053218.1 | BAC55019.1 | |||
AK155336.1 | BAE33202.1 | |||
AK178703.1 | ||||
AK183642.1 | ||||
AK205818.1 | ||||
AK211071.1 | ||||
AK216064.1 | ||||
BC026638.1 | AAH26638.1 | |||
BC031613.1 | AAH31613.1 | |||
CX205743.1 | ||||
CX206051.1 |
More Information
Set Membership: |
Intragenic Non-exonic |