Assay Details
Target Gene Details
Entrez Gene ID: | 217127 |
Gene Name: | K(lysine) acetyltransferase 7 |
Gene Aliases: |
Hbo1, Hboa, Myst2 |
Location: |
Chr.11:95271853-95310246 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat7 | NM_001195003.1 | 5 | 825 | NP_001181932.1 |
NM_001195004.1 | 5 | 825 | NP_001181933.1 | |
NM_177619.3 | 4 | 648 | NP_808287.1 | |
AK147968.1 | 4 | 552 | BAE28257.1 | |
AK169458.1 | 5 | 747 | ||
AK170389.1 | 5 | 772 | BAE41763.1 | |
AK177238.1 | 2 | 66 | ||
AK181521.1 | ||||
AK186978.1 | ||||
BC048904.1 | 4 | 603 | AAH48904.1 | |
BC057102.1 | 5 | 753 | AAH57102.1 | |
DQ076247.1 | 5 | 753 |
More Information
Set Membership: |
Intragenic Exonic |