Assay Details
Target Gene Details
Entrez Gene ID: | 14377 |
Gene Name: | glucose-6-phosphatase, catalytic |
Gene Aliases: |
AW107337, G6Pase, G6pt, Glc-6-Pase |
Location: |
Chr.11:101367730-101377903 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
G6pc | NM_008061.3 | NP_032087.2 | ||
AK050279.1 | BAC34162.1 | |||
AK052656.1 | BAC35084.1 | |||
BC013448.1 | AAH13448.1 | |||
U00445.1 | AAC52122.1 |
More Information
Set Membership: |
Intragenic Non-exonic |