Assay Details
Target Gene Details
Entrez Gene ID: | 13866 |
Gene Name: | v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) |
Gene Aliases: |
Erbb-2, HER-2, HER2, Neu, c-erbB2, c-neu, mKIAA3023 |
Location: |
Chr.11:98412484-98437716 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erbb2 | NM_001003817.1 | NP_001003817.1 | ||
AK031099.1 | ||||
AK083669.1 | ||||
AK129487.1 | BAC98297.1 | |||
AK142296.1 | ||||
BC046811.1 | AAH46811.1 | |||
BC053078.1 | AAH53078.1 |
More Information
Set Membership: |
Intragenic Non-exonic |