Assay Details
Target Gene Details
Entrez Gene ID: | 432572 |
Gene Name: | sperm antigen with calponin homology and coiled-coil domains 1 |
Gene Aliases: |
2810012G08Rik, B230396K10Rik, Cytsb |
Location: |
Chr.11:61956763-62223013 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Specc1 | NM_001029936.3 | 1 | 58 | NP_001025107.1 |
NM_001281818.1 | NP_001268747.1 | |||
AK046469.1 | BAC32744.2 | |||
AK050982.1 | 1 | 59 | BAC34484.2 | |
AK143663.1 | BAE25484.1 | |||
AK220465.1 | BAD90492.1 |