Assay Details
Target Gene Details
Entrez Gene ID: | 118454 |
Gene Name: | gap junction protein, gamma 2 |
Gene Aliases: |
B230382L12Rik, Cx47, Gja12 |
Location: |
Chr.11:59175564-59183213 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gjc2 | NM_080454.4 | NP_536702.3 | ||
NM_175452.4 | NP_780661.2 | |||
AK046421.1 | BAC32722.1 | |||
AK046609.1 | BAC32806.1 | |||
AY394498.1 | AAR05823.1 |
More Information
Set Membership: |
Intragenic Non-exonic |