Assay Details
Target Gene Details
Entrez Gene ID: | 14615 |
Gene Name: | gap junction protein, gamma 1 |
Gene Aliases: |
C130009G16Rik, Cnx45, Cx45, Gja-7, Gja7 |
Location: |
Chr.11:102799472-102819686 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gjc1 | NM_001159382.1 | NP_001152854.1 | ||
NM_001159383.1 | NP_001152855.1 | |||
NM_008122.3 | NP_032148.1 | |||
AK081351.1 | BAC38203.1 | |||
AK085224.1 | 1 | 2146 | ||
AK085236.1 | 1 | 2146 | ||
AY390396.1 | ||||
AY390397.1 | ||||
BC050840.1 | AAH50840.1 | |||
BC071230.1 | AAH71230.1 | |||
X63100.1 | CAA44814.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |