Assay Details
Target Gene Details
Entrez Gene ID: | 246746 |
Gene Name: | CD300 antigen like family member F |
Gene Aliases: |
CLIM1, CLM-1, CLM1, Digr2, F730004D16Rik, IREM1, IgSF13, LMIR3, Pigr3 |
Location: |
Chr.11:115116214-115134150 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd300lf | NM_001169153.1 | 6 | 879 | NP_001162624.1 |
NM_145634.3 | 6 | 858 | NP_663609.2 | |
AB292061.1 | 6 | 768 | ||
AF251703.1 | AAM19096.1 | |||
AK089313.1 | 6 | 879 | BAC40838.1 | |
AK154273.1 | 5 | 689 | BAE32481.1 | |
AK155367.1 | 6 | 856 | BAE33222.1 | |
AY214460.1 | 6 | 810 | AAO59487.1 | |
AY457047.1 | 6 | 768 | AAR27938.1 | |
BC057864.1 | 6 | 823 | AAH57864.1 | |
JX073136.1 | 5 | 724 |
Target Gene Details
Entrez Gene ID: | 58222 |
Gene Name: | RAB37, member RAS oncogene family |
Gene Aliases: |
B230331O03Rik, B230354I04Rik |
Location: |
Chr.11:115091203-115162773 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rab37 | NM_001163753.1 | NP_001157225.1 | ||
AK045995.1 | ||||
AK046260.1 | BAC32661.1 | |||
AK134576.1 | ||||
AK138293.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |