Assay Details
Target Gene Details
Entrez Gene ID: | 53413 |
Gene Name: | exocyst complex component 7 |
Gene Aliases: |
Exo70, sec70 |
Location: |
Chr.11:116287998-116306749 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Exoc7 | NM_001162872.1 | NP_001156344.1 | ||
NM_016857.2 | NP_058553.2 | |||
AF014461.1 | AAB69345.1 | |||
AK087601.1 | BAC39941.1 | |||
AK140450.1 | BAE24393.1 | |||
AK167769.1 | BAE39802.1 | |||
AK169928.1 | BAE41466.1 | |||
AK173094.1 | BAD32372.1 | |||
BC028927.1 | AAH28927.1 | |||
BC053710.1 | AAH53710.1 |
More Information
Set Membership: |
Intragenic Non-exonic |