Assay Details
Target Gene Details
Entrez Gene ID: | 12957 |
Gene Name: | crystallin, beta A1 |
Gene Aliases: |
BA3/A1, Cryb |
Location: |
Chr.11:77718732-77725781 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cryba1 | NM_009965.2 | 4 | 297 | NP_034095.1 |
AJ239052.1 | 4 | 297 | CAB52418.1 | |
AK084145.1 | 1 | 874 | BAC39126.1 | |
BC107342.2 | 4 | 297 | AAI07343.1 | |
J00378.1 | 1 | 70 | AAA51629.1 | |
V00728.1 | 2 | 189 | CAA24106.1 |