Assay Details
Target Gene Details
Entrez Gene ID: | 20308 |
Gene Name: | chemokine (C-C motif) ligand 9 |
Gene Aliases: |
CCF18, MRP-2, Scya10, Scya9 |
Location: |
Chr.11:83572917-83578636 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl9 | NM_011338.2 | NP_035468.1 | ||
AF128195.1 | AAF22536.1 | |||
AF128196.1 | AAF22537.1 | |||
AF128197.1 | AAF22538.1 | |||
AF128198.1 | AAF22539.1 | |||
AF128199.1 | AAF22540.1 | |||
AF128200.1 | AAF22541.1 | |||
AF128201.1 | AAF22542.1 | |||
AF128202.1 | AAF22543.1 | |||
AF128203.1 | AAF22544.1 | |||
AF128204.1 | AAF22545.1 | |||
AK151131.1 | BAE30139.1 | |||
AK151649.1 | BAE30578.1 | |||
AK154511.1 | BAE32640.1 | |||
AK154657.1 | BAE32747.1 | |||
AK155032.1 | BAE33004.1 | |||
AK155036.1 | BAE33005.1 | |||
BC139318.1 | ||||
BC145962.1 | ||||
BE626870.1 | ||||
CB950873.1 | ||||
U15209.1 | AAA92583.1 | |||
U19482.1 | AAB17120.1 | |||
U49513.1 | AAB02198.1 |
More Information
Set Membership: |
Intragenic Non-exonic |