Assay Details
Target Gene Details
Entrez Gene ID: | 20307 |
Gene Name: | chemokine (C-C motif) ligand 8 |
Gene Aliases: |
1810063B20Rik, AB023418, HC14, MCP-2, Mcp2, Scya8 |
Location: |
Chr.11:82115185-82116799 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl8 | NM_021443.3 | NP_067418.1 | ||
AB023418.1 | BAA75014.1 | |||
AK007942.1 | BAB25365.1 | |||
BC117101.1 | ||||
BC117103.1 | ||||
BY707942.1 |
More Information
Set Membership: |
Intragenic Non-exonic |