Assay Details
Target Gene Details
Entrez Gene ID: | 67291 |
Gene Name: | coiled-coil domain containing 137 |
Gene Aliases: |
3110023B02Rik |
Location: |
Chr.11:120458129-120464358 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc137 | NM_152807.3 | NP_690020.1 | ||
AK156682.1 | BAE33806.1 | |||
BC026662.1 | AAH26662.1 |