Assay Details
Target Gene Details
Entrez Gene ID: | 69131 |
Gene Name: | cyclin-dependent kinase 12 |
Gene Aliases: |
1810022J16Rik, AI646528, Crk7, Crkrs, D11Ertd752e, Pksc |
Location: |
Chr.11:98202326-98281101 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdk12 | NM_001109626.1 | 8 | 2762 | NP_001103096.1 |
NM_001109628.1 | 8 | 2762 | NP_001103098.1 | |
NM_026952.2 | 8 | 2762 | NP_081228.2 | |
AK049376.1 | 8 | 2408 | ||
AK129237.1 | 8 | 1901 | BAC98047.1 | |
AK137734.1 | 6 | 632 | ||
AY072295.1 | 8 | 2753 | AAL69526.1 | |
BC116645.1 | 8 | 2745 |
More Information
Set Membership: |
Intragenic Exonic |