Assay Details
Target Gene Details
Entrez Gene ID: | 20303 |
Gene Name: | chemokine (C-C motif) ligand 4 |
Gene Aliases: |
AT744.1, Act-2, MIP-1B, Mip1b, Scya4 |
Location: |
Chr.11:83648624-83664683 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl4 | NM_013652.2 | NP_038680.1 | ||
AF128218.1 | AAF22559.1 | |||
AF128219.1 | AAF22560.1 | |||
AK089326.1 | 1 | 887 | ||
AK155053.1 | 1 | 887 | ||
AK156948.1 | BAE33909.1 | |||
AK172472.1 | BAE43025.1 | |||
BC119257.1 | ||||
BC119259.1 | ||||
BY163868.1 | ||||
M23503.1 | AAA40148.1 | |||
M35590.1 | AAA39708.1 |
More Information
Set Membership: |
Intragenic Non-exonic |