Assay Details
Target Gene Details
Entrez Gene ID: | 20296 |
Gene Name: | chemokine (C-C motif) ligand 2 |
Gene Aliases: |
AI323594, HC11, JE, MCAF, MCP-1, MCP1, SMC-CF, Scya2, Sigje |
Location: |
Chr.11:82035577-82037452 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl2 | NM_011333.3 | NP_035463.1 | ||
AF065929.1 | AAF15379.1 | |||
AF065930.1 | AAF15380.1 | |||
AF065931.1 | AAF15381.1 | |||
AF065932.1 | AAF15382.1 | |||
AF065933.1 | AAF15383.1 | |||
AK132590.1 | BAE21250.1 | |||
AK150937.1 | BAE29972.1 | |||
AK151789.1 | BAE30690.1 | |||
AK153443.1 | BAE31998.1 | |||
AK153468.1 | BAE32019.1 | |||
AK153520.1 | BAE32062.1 | |||
BC055070.1 | AAH55070.1 | |||
BC145867.1 | ||||
BC145869.1 | ||||
CT010187.1 | CAJ18395.1 |
More Information
Set Membership: |
Intragenic Non-exonic |