Assay Details
Target Gene Details
Entrez Gene ID: | 20290 |
Gene Name: | chemokine (C-C motif) ligand 1 |
Gene Aliases: |
BF534335, I-309, Scya1, Tca-3 |
Location: |
Chr.11:82176666-82179884 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl1 | NM_011329.3 | 1 | 73 | NP_035459.1 |
AF065924.1 | 1 | 52 | AAF15374.1 | |
AF065925.1 | 1 | 52 | AAF15375.1 | |
AF065926.1 | 1 | 52 | AAF15376.1 | |
AF065927.1 | 1 | 52 | AAF15377.1 | |
AF065928.1 | 1 | 52 | AAF15378.1 | |
AK019040.1 | 1 | 73 | BAB31521.1 | |
BC120806.1 | 1 | 64 | ||
M17957.1 | 1 | 76 | AAB39499.1 | |
M23501.1 | 1 | 73 | AAA40147.1 |
More Information
Set Membership: |
Intragenic Exonic |