Assay Details
Target Gene Details
Entrez Gene ID: | 20293 |
Gene Name: | chemokine (C-C motif) ligand 12 |
Gene Aliases: |
MCP-5, Scya12 |
Location: |
Chr.11:82101845-82103399 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl12 | NM_011331.2 | NP_035461.2 | ||
AF065934.1 | AAF15384.1 | |||
AF065935.1 | AAF15385.1 | |||
AF065936.1 | AAF15386.1 | |||
AF065937.1 | AAF15387.1 | |||
AF065938.1 | AAF15388.1 | |||
AK012356.1 | BAB28185.1 | |||
BC027520.1 | AAH27520.1 | |||
U50712.1 | AAB50053.1 | |||
U66670.1 | AAB49424.1 |
More Information
Set Membership: |
Intragenic Non-exonic |