Assay Details
Target Gene Details
Entrez Gene ID: | 20292 |
Gene Name: | chemokine (C-C motif) ligand 11 |
Gene Aliases: |
Scya11, eotaxin |
Location: |
Chr.11:82057832-82062955 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl11 | NM_011330.3 | NP_035460.1 | ||
AF128205.1 | AAF22546.1 | |||
AF128206.1 | AAF22547.1 | |||
AF128207.1 | AAF22548.1 | |||
AF128208.1 | AAF22549.1 | |||
AF128209.1 | AAF22550.1 | |||
AK009307.1 | ||||
AK010146.1 | BAB26731.1 | |||
AW986913.1 | ||||
BC027521.1 | AAH27521.1 | |||
U26426.1 | AAC52256.1 | |||
U40672.1 | AAA99776.1 |
More Information
Set Membership: |
Intragenic Non-exonic |