Assay Details
Target Gene Details
Entrez Gene ID: | 13591 |
Gene Name: | early B cell factor 1 |
Gene Aliases: |
Ebf, O/E-1, OE-1, Olf-1, Olf1 |
Location: |
Chr.11:44618100-45008096 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ebf1 | NM_001290709.1 | 1 | 85 | NP_001277638.1 |
NM_001290710.1 | 1 | 85 | NP_001277639.1 | |
NM_001290711.1 | 1 | 85 | NP_001277640.1 | |
NM_007897.3 | 1 | 85 | NP_031923.1 | |
AK031239.1 | 1 | 80 | BAC27313.1 | |
AK035781.1 | BAC29185.1 | |||
AK051438.1 | 1 | 84 | BAE43347.1 | |
AK137389.1 | ||||
AK139021.1 | 1 | 83 | BAE23864.1 | |
AK141175.1 | 1 | 85 | BAE24573.1 | |
BC139362.1 | 1 | 46 | ||
BC139364.1 | 1 | 46 | ||
L12147.1 | 1 | 51 | AAA37533.1 |
More Information
Set Membership: |
Intragenic Exonic |