Assay Details
Target Gene Details
Entrez Gene ID: | 18612 |
Gene Name: | ets variant 4 |
Gene Aliases: |
AW414408, Pea-3, Pea3 |
Location: |
Chr.11:101769742-101785310 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Etv4 | NM_008815.2 | NP_032841.2 | ||
AK144590.1 | BAE25952.1 | |||
AK144683.1 | ||||
AK166136.1 | ||||
DQ832277.1 |
More Information
Set Membership: |
Intragenic Non-exonic |