Assay Details
Target Gene Details
Entrez Gene ID: | 210004 |
Gene Name: | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1 |
Gene Aliases: |
6030413G23Rik |
Location: |
Chr.11:121616197-121673160 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
B3gntl1 | NM_178664.5 | NP_848779.2 | ||
AK031361.1 | BAC27366.1 | |||
AK156318.1 | BAE33671.1 | |||
BC022739.1 | AAH22739.1 | |||
BC088993.1 | AAH88993.1 | |||
BC125626.1 | ||||
BC132193.1 | ||||
BC145166.1 |
More Information
Set Membership: |
Intragenic Non-exonic |