Assay Details
Target Gene Details
Entrez Gene ID: | 56697 |
Gene Name: | A kinase (PRKA) anchor protein 10 |
Gene Aliases: |
1500031L16Rik, B130049N18Rik, D-AKAP-2, D-AKAP2, PRKA10 |
Location: |
Chr.11:61871307-61930307 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Akap10 | NM_019921.3 | 10 | 1738 | NP_064305.2 |
AF021833.1 | 7 | 1092 | AAC61898.1 | |
AK049399.1 | 10 | 1707 | BAC33735.1 | |
AK051044.1 | 8 | 1373 | ||
AK083255.1 | 10 | 1707 | ||
AK162966.1 | 8 | 1530 | BAE37139.1 | |
BC054105.1 | 7 | 1041 | AAH54105.1 |
More Information
Set Membership: |
Intragenic Exonic |