Assay Details
Target Gene Details
Entrez Gene ID: | 11370 |
Gene Name: | acyl-Coenzyme A dehydrogenase, very long chain |
Gene Aliases: |
vlcad |
Location: |
Chr.11:70010183-70015428 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acadvl | NM_017366.3 | NP_059062.1 | ||
AF017176.1 | AAC31642.1 | |||
AK076037.1 | ||||
AK146334.1 | BAE27089.1 | |||
AK211026.1 | ||||
BC026559.1 | AAH26559.1 | |||
Z71189.1 | CAA94919.2 |
More Information
Set Membership: |
Intragenic Non-exonic |