Assay Details
Target Gene Details
Entrez Gene ID: | 11370 |
Gene Name: | acyl-Coenzyme A dehydrogenase, very long chain |
Gene Aliases: |
vlcad |
Location: |
Chr.11:70010183-70015428 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acadvl | NM_017366.3 | 11 | 1240 | NP_059062.1 |
AF017176.1 | 8 | 888 | AAC31642.1 | |
AK076037.1 | 11 | 1201 | ||
AK146334.1 | 6 | 713 | BAE27089.1 | |
BC026559.1 | 11 | 1219 | AAH26559.1 | |
U41497.1 | 2 | 122 | AAA85185.1 | |
Z71189.1 | 11 | 1161 | CAA94919.2 |
More Information
Set Membership: |
Intragenic Exonic |