Assay Details
Target Gene Details
Entrez Gene ID: | 56208 |
Gene Name: | beclin 1, autophagy related |
Gene Aliases: |
4921513J16Rik, 5430417M23Rik, Atg6 |
Location: |
Chr.11:101288262-101302288 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Becn1 | NM_019584.3 | NP_062530.2 | ||
AF077302.2 | AAC68654.2 | |||
AK028851.1 | ||||
AK031501.1 | ||||
AK050329.1 | BAC34192.1 | |||
AK077392.1 | ||||
AK083800.1 | BAC39021.1 | |||
AK151531.1 | ||||
AK177644.1 | ||||
AK178460.1 | ||||
AK178637.1 | ||||
BC005770.1 | AAH05770.1 |
More Information
Set Membership: |
Intragenic Non-exonic |