Assay Details
Target Gene Details
Entrez Gene ID: | 68876 |
Gene Name: | XRCC6 binding protein 1 |
Gene Aliases: |
1110068E08Rik, 2410012H02Rik |
Location: |
Chr.10:126868427-126901711 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Xrcc6bp1 | NM_001159559.1 | 4 | 460 | NP_001153031.1 |
NM_026858.3 | 4 | 460 | NP_081134.2 | |
AK004405.1 | 4 | 440 | ||
AK010471.1 | 4 | 443 | BAB26964.1 | |
AK039002.1 | 4 | 441 | ||
BC030630.1 | 4 | 422 | AAH30630.1 | |
BG801921.1 | ||||
CB203044.1 | 1 | 75 |
More Information
Set Membership: |
Intragenic Exonic |