Assay Details
Target Gene Details
Entrez Gene ID: | 23801 |
Gene Name: | arachidonate lipoxygenase 3 |
Gene Aliases: |
e-LOX-3, eLOX-3 |
Location: |
Chr.11:69125674-69149115 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aloxe3 | NM_011786.2 | 1 | 119 | NP_035916.2 |
AK141003.1 | 1 | 282 | ||
BC116258.1 | 1 | 118 | ||
BC116259.1 | 1 | 118 | ||
Y14695.1 | 1 | 119 | CAB46101.1 |
More Information
Set Membership: |
Intragenic Exonic |