Assay Details
Target Gene Details
Entrez Gene ID: | 66753 |
Gene Name: | endoplasmic reticulum lectin 1 |
Gene Aliases: |
4933407N01Rik |
Location: |
Chr.11:30865632-30954173 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erlec1 | NM_025745.3 | 9 | 976 | NP_080021.3 |
AK002471.1 | 10 | 967 | BAB22125.1 | |
AK077120.1 | 9 | 969 | BAC36623.1 | |
AK133874.1 | 9 | 977 | BAE21904.1 | |
BC018468.1 | 9 | 986 | AAH18468.1 |