Assay Details
Target Gene Details
Entrez Gene ID: | 11684 |
Gene Name: | arachidonate 12-lipoxygenase |
Gene Aliases: |
9930022G08Rik, Alox12p, P-12LO |
Location: |
Chr.11:70241455-70255386 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Alox12 | NM_007440.4 | 5 | 664 | NP_031466.2 |
AK036898.1 | 5 | 662 | BAC29629.1 | |
AK036960.1 | 5 | 620 | ||
AK079232.1 | 5 | 665 | BAC37583.1 | |
AK087724.1 | 5 | 665 | BAC39981.1 | |
AK172216.1 | 5 | 662 | BAE42887.1 | |
AK172488.1 | 5 | 665 | BAE43031.1 | |
BC152329.1 | 5 | 669 | ||
BC152403.1 | 5 | 669 | ||
S77511.1 | 3 | 221 | AAB34667.1 | |
S80446.1 | 5 | 676 | AAB36013.1 | |
U04334.1 | 5 | 595 | AAA20659.1 |
More Information
Set Membership: |
Intragenic Exonic |