Assay Details
Target Gene Details
Entrez Gene ID: | 22361 |
Gene Name: | vanin 1 |
Gene Aliases: |
V-1 |
Location: |
Chr.10:23894688-23905344 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vnn1 | NM_011704.3 | NP_035834.2 | ||
AJ132098.1 | CAA10567.1 | |||
AK002773.1 | BAB22347.1 | |||
AK145984.1 | BAE26806.1 | |||
AK167427.1 | BAE39515.1 | |||
BB838710.1 | ||||
BC019203.1 | AAH19203.1 |
More Information
Set Membership: |
Intragenic Non-exonic |