Assay Details
Target Gene Details
Entrez Gene ID: | 70419 |
Gene Name: | RIKEN cDNA 2810408A11 gene |
Gene Aliases: |
1700019L01Rik, 4930470J18Rik |
Location: |
Chr.11:69897358-69901721 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
2810408A11Rik | NM_027419.3 | NP_081695.2 | ||
AK013042.1 | BAB28617.1 | |||
AK015537.1 | ||||
AK051029.1 | ||||
AK160144.1 | ||||
AK171674.1 | BAE42603.1 | |||
BC005538.1 | ||||
BC051546.1 | ||||
BC069874.1 | AAH69874.1 |
More Information
Set Membership: |
Intragenic Non-exonic |