Assay Details
Target Gene Details
Entrez Gene ID: | 14422 |
Gene Name: | beta-1,4-N-acetyl-galactosaminyl transferase 2 |
Gene Aliases: |
AI593864, Dlb-1, Dlb1, Galg2, Galgt2, Ggm3, Igf2bp1 |
Location: |
Chr.11:95863559-95914920 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
B4galnt2 | NM_008081.3 | NP_032107.1 | ||
BC139165.1 | ||||
BC139166.1 | ||||
L30104.1 | AAA39802.1 |