Assay Details
Target Gene Details
Entrez Gene ID: | 216345 |
Gene Name: | zinc finger, C3H1-type containing |
Gene Aliases: |
BC033596, Ccdc131, Psrc2 |
Location: |
Chr.10:115384920-115432772 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfc3h1 | NM_001033261.2 | 9 | 2222 | NP_001028433.2 |
AK145652.1 | 9 | 2224 | ||
AK172969.1 | 7 | 922 | BAD32247.1 | |
BC139124.1 | 9 | 2079 |