Assay Details
Target Gene Details
Entrez Gene ID: | 93759 |
Gene Name: | sirtuin 1 |
Gene Aliases: |
AA673258, SIR2L1, Sir2, Sir2a, Sir2alpha |
Location: |
Chr.10:63319005-63339061 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sirt1 | NM_001159589.1 | NP_001153061.1 | ||
NM_019812.2 | NP_062786.1 | |||
AF214646.1 | AAF24983.1 | |||
AK144203.1 | BAE25766.1 | |||
AY377984.1 | AAR23928.1 | |||
BC152314.1 |