Assay Details
Target Gene Details
Entrez Gene ID: | 268281 |
Gene Name: | SNF2 histone linker PHD RING helicase |
Gene Aliases: |
2610103K11Rik, AA450458, AU024614, BC006883, D230017O13Rik, E130018M05 |
Location: |
Chr.10:11149321-11217595 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shprh | NM_001077707.1 | 21 | 4056 | NP_001071175.1 |
NM_001284354.1 | 21 | 4056 | NP_001271283.1 | |
NM_172937.3 | 21 | 4056 | NP_766525.3 | |
AK082160.1 | 21 | 4067 | BAC38428.1 | |
AK145015.1 | BAE26184.1 | |||
AY162264.1 | 21 | 3962 | AAO26654.1 | |
AY162266.1 | 21 | 3962 | AAO26656.1 | |
BC006883.1 | 5 | 583 | AAH06883.1 | |
BC055003.1 | 21 | 4027 | AAH55003.1 |
More Information
Set Membership: |
Intragenic Exonic |