Assay Details
Target Gene Details
Entrez Gene ID: | 268281 |
Gene Name: | SNF2 histone linker PHD RING helicase |
Gene Aliases: |
2610103K11Rik, AA450458, AU024614, BC006883, D230017O13Rik, E130018M05 |
Location: |
Chr.10:11149321-11217595 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shprh | NM_001077707.1 | NP_001071175.1 | ||
NM_001284354.1 | NP_001271283.1 | |||
NM_172937.3 | NP_766525.3 | |||
AK053448.1 | BAC35389.1 | |||
AK082160.1 | BAC38428.1 | |||
AK145015.1 | BAE26184.1 | |||
AY162264.1 | AAO26654.1 | |||
AY162266.1 | AAO26656.1 | |||
BC055003.1 | AAH55003.1 |
More Information
Set Membership: |
Intragenic Non-exonic |