Assay Details
Target Gene Details
Entrez Gene ID: | 76142 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 14c |
Gene Aliases: |
6330514J04Rik, KEPI |
Location: |
Chr.10:3366071-3464979 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r14c | NM_133485.2 | NP_597844.1 | ||
AF407167.2 | AAL83508.1 | |||
AK085997.1 | BAC39589.1 | |||
BC055717.1 | AAH55717.1 |
More Information
Set Membership: |
Intragenic Non-exonic |