Assay Details
Target Gene Details
Entrez Gene ID: | 319468 |
Gene Name: | protein phosphatase 1H (PP2C domain containing) |
Gene Aliases: |
A430075L18Rik, ARHCL1, C030002B11Rik |
Location: |
Chr.10:122678106-122945795 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppm1h | NM_001110218.1 | NP_001103688.1 | ||
NM_176919.4 | NP_795893.2 | |||
AK040194.1 | BAC30536.1 | |||
AK040207.1 | BAE43308.1 | |||
AK051434.1 | ||||
AK077422.1 | ||||
AK134804.1 | BAE22292.1 | |||
AK150309.1 | BAE29457.1 | |||
AK220260.1 | BAD90185.1 | |||
BC052910.1 | AAH52910.1 |
More Information
Set Membership: |
Intragenic Non-exonic |