Assay Details
Target Gene Details
Entrez Gene ID: | 20598 |
Gene Name: | sphingomyelin phosphodiesterase 2, neutral |
Gene Aliases: |
AW108287, nSMase, nSMase1 |
Location: |
Chr.10:41487172-41490340 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smpd2 | NM_009213.2 | 4 | 579 | NP_033239.1 |
AJ222800.1 | 4 | 580 | CAA10994.1 | |
AK075616.1 | 4 | 589 | BAC35861.1 | |
AK088962.1 | 2 | 1242 | ||
BC010978.1 | 4 | 589 | AAH10978.1 |
More Information
Set Membership: |
Intragenic Exonic |