Assay Details
Target Gene Details
Entrez Gene ID: | 66118 |
Gene Name: | SAP domain containing ribonucleoprotein |
Gene Aliases: |
1110005A23Rik, Cip29 |
Location: |
Chr.10:128821766-128877638 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sarnp | NM_025364.2 | 1 | 62 | NP_079640.1 |
AW824990.1 | 1 | 69 |