Assay Details
Target Gene Details
Entrez Gene ID: | 71137 |
Gene Name: | regulatory factor X, 4 (influences HLA class II expression) |
Gene Aliases: |
4933412G19Rik, NYD-sp10 |
Location: |
Chr.10:84756048-84906538 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rfx4 | NM_001024918.1 | NP_001020089.1 | ||
NM_027689.3 | NP_081965.2 | |||
AB086957.1 | BAD07408.1 | |||
AB089184.1 | BAD07409.1 | |||
AK034131.1 | BAC28598.1 | |||
AK132673.1 | BAE21294.1 | |||
AY102010.2 | AAM52485.2 | |||
AY342003.1 | AAQ17042.1 | |||
BC138719.1 | ||||
BC138720.1 |
More Information
Set Membership: |
Intragenic Non-exonic |