Assay Details
Target Gene Details
Entrez Gene ID: | 270685 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like |
Gene Aliases: |
2410004L15Rik, AI647056, Fthfsdc1 |
Location: |
Chr.10:3970441-4167081 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mthfd1l | NM_001170785.1 | NP_001164256.1 | ||
NM_001170786.1 | NP_001164257.1 | |||
NM_172308.4 | NP_758512.3 | |||
AK028211.1 | BAE20438.1 | |||
AK036284.1 | BAE20500.1 | |||
AK038579.1 | BAC30053.1 | |||
AK075931.1 | ||||
AK160025.1 | BAE35568.1 | |||
AK181581.1 | ||||
AK185610.1 | ||||
BC030437.1 | AAH30437.1 | |||
BC049936.1 | AAH49936.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |