Assay Details
Target Gene Details
Entrez Gene ID: | 14816 |
Gene Name: | glutamate receptor, metabotropic 1 |
Gene Aliases: |
4930455H15Rik, ENSMUSG00000075319, Gm10828, Gprc1a, mGluR1, nmf373, rcw, wobl |
Location: |
Chr.10:10686059-11082377 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grm1 | NM_001114333.2 | 3 | 1143 | NP_001107805.1 |
NM_016976.3 | 3 | 1143 | NP_058672.1 | |
AF320126.2 | 3 | 1168 | AAG41991.2 | |
BC067057.1 | 3 | 1117 | AAH67057.1 | |
BC079566.1 | 2 | 1160 | AAH79566.1 |
More Information
Set Membership: |
Intragenic Exonic |