Assay Details
Target Gene Details
Entrez Gene ID: | 237310 |
Gene Name: | interleukin 22 receptor, alpha 2 |
Gene Aliases: |
CRF2-10, CRF2-s1, CRF2X, Il-22bp |
Location: |
Chr.10:19622028-19634681 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il22ra2 | NM_178258.5 | NP_839989.2 | ||
AF493604.1 | AAP13730.1 | |||
AJ555484.1 | CAD88474.1 | |||
AK040641.1 | BAE20584.1 | |||
AK138913.1 | BAE23815.1 | |||
AK139024.1 | BAE23866.1 | |||
BC124998.1 | ||||
BC126872.1 |
More Information
Set Membership: |
Intragenic Non-exonic |