Assay Details
Target Gene Details
Entrez Gene ID: | 50929 |
Gene Name: | interleukin 22 |
Gene Aliases: |
IL-22, IL-22a, ILTIFa, Iltif |
Location: |
Chr.10:118204586-118210047 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il22 |