Assay Details
Target Gene Details
Entrez Gene ID: | 22781 |
Gene Name: | IKAROS family zinc finger 4 |
Gene Aliases: |
A630026H08Rik, Eos, Zfpn1a4, Znfn1a4 |
Location: |
Chr.10:128630839-128669358 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ikzf4 | NM_011772.2 | NP_035902.2 | ||
AB017615.1 | BAA36213.1 | |||
AK089522.1 | BAC40912.1 | |||
AK170453.1 | BAE41808.1 | |||
BC141286.1 |
More Information
Set Membership: |
Intragenic Non-exonic |